Mouse MMP2 ELISA Kit
Catalog # ABCAAB100730-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse MMP2 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse MMP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MMP2 in Mouse in Plasma, Cell culture supernatant, Serum samples.