ContactUsLinkComponent
Human Adiponectin ELISA Kit
Human Adiponectin ELISA Kit
Catalog # ABCAAB108786-96
Supplier:  Abcam
CAS Number:  
undefined
Human Adiponectin ELISA Kit
Catalog # ABCAAB108786-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Adiponectin ELISA kit
Specifications
About this item
Human Adiponectin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Adiponectin in Human in Saliva, urine, plasma, cell culture supernatant, milk, serum, Cerebral Spinal Fluid samples.