Human Peroxiredoxin 2/PRP ELISA Kit
Catalog # ABCAAB277456-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Peroxiredoxin 2/PRP ELISA kit
Specifications
About this item
Human Peroxiredoxin 2/PRP ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Peroxiredoxin 2/PRP in Human in plasma, cell culture supernatant, serum samples.