ContactUsLinkComponent
C-Peptide ELISA Kit
C-Peptide ELISA Kit
Catalog # ABCAAB178641-96
Supplier:  Abcam
CAS Number:  
undefined
C-Peptide ELISA Kit
Catalog # ABCAAB178641-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    C-Peptide ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
C-Peptide ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of C-Peptide in Human in Citrate plasma, Heparin Plasma, serum samples.
Certifications: RUO