Bovine IP-10 ELISA Kit (CXCL10)
Catalog # ABCAAB273198-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Bovine IP-10 ELISA Kit (CXCL10)
- Tests per kit:96 tests
Specifications
About this item
Bovine IP-10 ELISA Kit (CXCL10) is a Sandwich (quantitative) ELISA kit for the measurement of Bovine IP-10 (CXCL10) in Cow in Plasma, Cell culture supernatant, Serum samples.