Rat Clusterin ELISA Kit
Catalog # ABCAAB231934-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Clusterin ELISA kit
Specifications
About this item
Rat Clusterin ELISA kit is a single-wash 90-min Simplestep used to quantify Rat Clusterin with a sensitivity of 0,63 µg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.