Mouse PD-L2 ELISA Kit
Catalog # ABCAAB283880-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse PD-L2 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse PD-L2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse PD-L2 with a sensitivity of 1,03 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.