Human Anti-Toxoplasma gondii IgM ELISA Kit
ABCAAB108778-96
: Abcam
:
- Description:Human Anti-Toxoplasma gondii IgM ELISA kit
Human Anti-Toxoplasma gondii IgM ELISA kit is a Sandwich (qualitative) ELISA kit for the measurement of Human Anti-Toxoplasma gondii IgM in Human in Citrate plasma, Heparin plasma, serum samples.