Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Human/Mouse Met (c-Met) Antibody Pair - BSA and Azide free
Human/Mouse Met (c-Met) Antibody Pair - BSA and Azide free
Human/Mouse Met (c-Met) Antibody Pair - BSA and Azide free
Catalog # ABCAAB253438-10X96
Supplier:
Abcam
CAS Number:
Expand image
Human/Mouse Met (c-Met) Antibody Pair - BSA and Azide free
Catalog # ABCAAB253438-10X96
Supplier:
Abcam
CAS Number:
Specifications
Description:
Human/Mouse Met (c-Met) antibody pair - BSA and Azide free
Tests per kit:
10×96 tests
Specifications
About this item
Product Details
Product Details