Mouse IGFBP3 ELISA Kit
Catalog # ABCAAB100692-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IGFBP3 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse IGFBP3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IGFBP3 in Mouse in Plasma, Cell culture supernatant, Serum samples.