Human IL-10 ELISA Kit (Interleukin-10)
Supplier: Abcam
New Product
Certificates
About this item
Human IL-10 ELISA kit (Interleukin-10) is a single-wash, 90-minute SimpleStep ELISA® assay designed for the quantification of human IL-10 with a sensitivity of 1,4 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric Sandwich ELISA - 450 nm readout; works on any plate reader. Different formats for different needs: 384-well for higher throughput - Broad sample compatibility - works with serum, plasma, and cell culture supernatants.
Specifications
- Storage temperature:+4 ℃