Human Complement C8 ELISA Kit
Catalog # ABCAAB137971-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Complement C8
- Description:Human Complement C8 ELISA kit
- Sample type:Plasma, serum, saliva, milk, cerebral spinal fluid
- Detection method:Colorimetric
- Time to Results:4 h 0 min
- Detection range:49 - 106 µg/ml
- Storage temperature:4 °C
- Sensitivity:0,24 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human Complement C8 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C8 in Human in plasma, Serum, Saliva, Milk, Cerebral Spinal Fluid samples.