Rat TIM 1 ELISA Kit (KIM-1)
Catalog # ABCAAB223858-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat TIM 1 ELISA kit (KIM-1)
- Tests per kit:96 tests
Specifications
About this item
Rat TIM 1 ELISA Kit (KIM-1) is a single-wash 90-min Simplestep used to quantify Rat TIM 1 (KIM-1) with a sensitivity of 2,33 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 5 citations.