Mouse IgG2a ELISA Kit
ABCAAB133046-96
: Abcam
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IgG2a
- Description:Mouse IgG2a ELISA kit
- Sample type:Serum, tissue culture media
- Detection method:Colorimetric
- Detection range:7,81 - 250 ng/ml
- Storage temperature:4 °C
- Sensitivity:47,3 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Mouse IgG2a ELISA kit ab133046 is used to quantify Mouse IgG2a with a sensitivity of 47.3 pg/ml. Sandwich ELISA kit.