Mouse Surfactant protein D / SP-D ELISA Kit
ABCAAB213890-96
: Abcam
:
- Description:Mouse Surfactant protein D / SP-D ELISA Kit
- Tests per kit:96 tests
Mouse Surfactant protein D / SP-D ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Surfactant protein D / SP-D in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.