Rat Alpha fetoprotein (AFP) Antibody Pair - BSA and Azide free
Catalog # ABCAAB253704-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Alpha fetoprotein (AFP) antibody pair - BSA and Azide free
- Tests per kit:10×96 tests
Specifications
About this item
Rat Alpha fetoprotein (AFP) Antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Rat Alpha fetoprotein (AFP).