Mouse/Rat IgM ELISA Kit
Catalog # ABCAAB215085-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:IgM
- Description:Mouse/Rat IgM ELISA kit
- Sample type:Heparin plasma, citrate plasma, cell culture supernatant, serum, edta plasma
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Detection range:1,56 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:385 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse/Rat IgM ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse/Rat IgM with a sensitivity of 385 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Cited in over 5 citations.