Mouse TNFRSF17 ELISA Kit (BCMA)
Catalog # ABCAAB213893-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse TNFRSF17 ELISA kit (BCMA)
- Tests per kit:96 tests
Specifications
About this item
Mouse TNFRSF17 ELISA kit (BCMA) is a sandwich (quantitative) ELISA kit for the measurement of Mouse TNFRSF17 (BCMA) in Mouse in Cell Lysate, cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.