Human GC Globulin ELISA Kit (Vitamin D Binding Protein)
Katalogové číslo ABCAAB108853-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:Vitamin D Binding protein
- Description:Human GC Globulin ELISA kit (Vitamin D Binding Protein)
- Sample type:Plasma, serum
- Detection method:Colorimetric
- Time to Results:3 h 0 min
- Detection range:0,098 - 100 µg/ml
- Storage temperature:4 °C
- Sensitivity:0,06 µg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human GC Globulin ELISA kit (Vitamin D Binding Protein) is a Competitive ELISA kit for the measurement of Human GC Globulin (Vitamin D Binding Protein) in Human in plasma, Serum samples.