ContactUsLinkComponent
Human VCAM-1 ELISA Set (without plates)
Human VCAM-1 ELISA Set (without plates)
Catalog # ABCAAB47355-480
Supplier:  Abcam
CAS Number:  
undefined
Human VCAM-1 ELISA Set (without plates)
Catalog # ABCAAB47355-480
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human VCAM-1 ELISA Set (without plates)
  • Seq:
    1
Specifications
About this item
Human VCAM-1 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human VCAM-1 ELISA Set (without plates) in Human in plasma, cell culture supernatant, serum samples.