Human Soluble TNF Receptor I ELISA Kit
Catalog # ABCAAB100642-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Soluble TNF Receptor I ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human Soluble TNF Receptor I ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Soluble TNF Receptor I in Human in Plasma, Cell culture supernatant, Serum samples.