8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA Kit
ABCAAB285254-1X96
: Abcam
:
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:8-hydroxy 2 deoxyguanosine
- Description:8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA kit
- Detection method:Colorimetric
- Detection range:1,563 - 100 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,94 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA kit is a Competitive ELISA kit for the measurement of 8-hydroxy-2'-deoxyguanosine (8-OHdG) in plasma, Tissue Homogenate, Serum, Other biological fluids samples.