Monkey IP-10 (CXCL10) ELISA Kit
Catalog # ABCAAB314607-96TST
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Monkey IP-10 (CXCL10) ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Monkey IP-10 (CXCL10) ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey IP-10 (CXCL10) with a sensitivity of 1,3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF).