Mouse RGMA ELISA Kit
Catalog # ABCAAB243687-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse RGMA ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse RGMA ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse RGMA with a sensitivity of 90 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.