Human IL-17 ELISA Set (without plates)
Catalog # ABCAAB83707-480
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-17 ELISA Set (without plates)
- Seq:1
- Tests per kit:5×96 tests
Specifications
About this item
Human IL-17 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-17 ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.