Human Kallikrein 6/KLK6 ELISA Kit
ABCAAB287163-1X96
: Abcam
:
- Description:Human Kallikrein 6/KLK6 ELISA kit
- Tests per kit:96 tests
Human Kallikrein 6/KLK6 ELISA kit is a single-wash 90-min Simplestep used to quantify Human Kallikrein 6/KLK6 with a sensitivity of 15,21 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.