anti-Zika virus IgM ELISA kit (mu-capture)
Catalog # ABCAAB213327-96
Supplier: Abcam
CAS Number:
Specifications
- Description:anti-Zika virus IgM ELISA kit (mu-capture)
- Tests per kit:96 tests
Specifications
About this item
anti-Zika virus IgM ELISA kit (μ-capture) is a Sandwich (qualitative) ELISA kit for the measurement of anti-Zika virus IgM (μ-capture) in Human in Citrate plasma, Heparin Plasma, Serum samples.