Mouse SAA ELISA Kit
Catalog # ABCAAB157723-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse SAA ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse SAA ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SAA in Mouse in Plasma, Serum samples.