Human Tissue Factor ELISA Kit
Catalog # ABCAAB220653-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Tissue Factor ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Tissue Factor ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Tissue Factor with a sensitivity of 3,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 10 citations.