Order Entry
Czech Republic
ContactUsLinkComponent
Human Fas Ligand ELISPOT Set
Human Fas Ligand ELISPOT Set
Catalog # ABCAAB46577-480
Supplier:  Abcam
CAS Number:  
undefined
Human Fas Ligand ELISPOT Set
Catalog # ABCAAB46577-480
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Fas Ligand ELISPOT set
  • Seq:
    1
  • Species reactivity:
    Human
  • Sample type:
    Suspension cells
  • Antigen name:
    Fas Ligand
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Product type:
    Uncoated
  • Tests per kit:
    5×96 tests

Specifications

About this item

Human Fas Ligand ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Fas Ligand production in a single cell suspension.