Human MCP-1 ELISA Kit
Supplier: Abcam
New Product
Certificates
About this item
Human MCP-1 ELISA kit is a single-wash 90-min Simplestep used to quantify Human MCP-1 with a sensitivity of 1,26 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Validated on a number of sample types including cerebrospinal fluid (CSF) - Design your own immunoassay: we also offer the conjugation-ready antibody pair.
Certifications: RUO
Frequently Bought Together