Order Entry
Czech Republic
ContactUsLinkComponent
Human C8a (Complement C8a) ELISA Kit
Human C8a (Complement C8a) ELISA Kit
Catalog # ABCAAB288173-1X96
Supplier:  Abcam
CAS Number:  
undefined
Human C8a (Complement C8a) ELISA Kit
Catalog # ABCAAB288173-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Complement C8a
  • Description:
    Human C8a (Complement C8a) ELISA kit
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    85,938 - 5500 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    40,141 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human C8a (Complement C8a) ELISA kit is a single-wash 90-min Simplestep used to quantify Human C8a (Complement C8a) with a sensitivity of 40.141 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.