ContactUsLinkComponent
Human Vimentin Profiling ELISA Kit
Human Vimentin Profiling ELISA Kit
Catalog # ABCAAB173190-96
Supplier:  Abcam
CAS Number:  
undefined
Human Vimentin Profiling ELISA Kit
Catalog # ABCAAB173190-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Vimentin Profiling ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
Human Vimentin Profiling ELISA Kit is a Quantitative ELISA kit for the measurement of Human Vimentin Profiling in Human in Tissue Lysate, Suspension cells, Tissue Homogenate, Adherent cells samples.