Mouse S100A4 ELISA Kit - Intracellular
Catalog # ABCAAB309319-96TST
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse S100A4 ELISA Kit - Intracellular
- Tests per kit:96 tests
Specifications
About this item
Mouse S100A4 ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Mouse S100A4 with a sensitivity of 5,7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.