Mouse Cardiac Troponin I Antibody Pair - BSA and Azide Free
Catalog # ABCAAB321920-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Cardiac Troponin I
- Description:Mouse Cardiac Troponin I antibody pair - BSA and azide free
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Mouse Cardiac Troponin I antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse Cardiac Troponin I.