Prostaglandin E2 ELISA Kit - Intracellular
: Abcam
New Product
Prostaglandin E2 ELISA Kit - Intracellular is a single-wash 90-min Simplestep used to quantify Prostaglandin E2 with a sensitivity of 6,88 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.
Frequently Bought Together