ContactUsLinkComponent
Human beta Nerve Growth Factor ELISA Kit
Human beta Nerve Growth Factor ELISA Kit
Catalog # ABCAAB193760-96
Supplier:  Abcam
CAS Number:  
undefined
Human beta Nerve Growth Factor ELISA Kit
Catalog # ABCAAB193760-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human beta Nerve Growth Factor ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Human beta Nerve Growth Factor ELISA Kit is a single-wash 90-min Simplestep used to quantify Human beta Nerve Growth Factor with a sensitivity of 6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 5 citations.