Assembly Gloves, profi ergo ENB20, 60148
Supplier: UVEX
Certificates
About this item
The thick special NBR coating of the uvex profi ergo is resistant to oil and grease. The high-quality cotton interlock lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate.
- Cotton interlock safety glove for universal use
- Palm and whole back of the hand with special NBR coating (nitrile rubber)
- Excellent wearer comfort with the high water vapour absorption of the cotton lining
The safety gloves in the uvex profi ergo series are characterised by very good grip in damp, wet and oily areas, good tactile feel, ergonomic fit, high flexibility and particularly good wearer comfort due to the high moisture absorption properties of the cotton lining. This glove is also suitable for operating touch displays. Free of harmful substances in line with OEKO-Tex® standard 100.
Specifications
- Catalog No:
- UVEX6014802
- UVEX6014803
- UVEX6014801
- UVEX6014804
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020
- EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020
- EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020
- EN 420:2003 + A1:2009/EN 388:2016 + A1:2018/EN 407:2020