Anti-Interferon alpha 2 Rabbit Monoclonal Antibody [clone: EPR19074-55]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR19074-55] to Interferon alpha 2 - BSA and Azide free (Detector).
Type: Primary
Antigen: Interferon alpha 2
Clonality: Monoclonal
Clone: EPR19074-55
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together