Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-AIDA Rabbit Monoclonal Antibody [clone: EPR20903-53]
Anti-AIDA Rabbit Monoclonal Antibody [clone: EPR20903-53]
Catalog # ABCAAB234419-1ML
Supplier:  Abcam
New Product
undefined
Anti-AIDA Rabbit Monoclonal Antibody [clone: EPR20903-53]
Catalog # ABCAAB234419-1ML
Supplier:  Abcam
New Product

Specifications

  • Sekvence:
    2070
  • Rozměry protilátky:
    1 ml
  • Typ balení:
    Vial

Specifications

About this item

Rabbit monoclonal [EPR20903-53] to AIDA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AIDA
Clonality: Monoclonal
Clone: EPR20903-53
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat