Anti-SAP30 Rabbit Polyclonal Antibody
ABCAAB231804-50
: Abcam
- Antibody type:Primary
- Antigen name:Histone deacetylase complex subunit SAP30
- Antigen symbol:SAP30
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- ChIP:Yes
- Form:Liquid
- Antigen synonyms:30 kDa Sin3-associated polypeptide|Sin3 corepressor complex subunit SAP30|Sin3-associated polypeptide p30
- Molecular weight:23306 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human Histone deacetylase complex subunit SAP30.
- Tested applications:ChIP-seq
- Purification:Protein G Affinity purification
- Size:50 µg
- Pack type:Vial
Rabbit polyclonal to SAP30.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SAP30
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human