Anti-MCPyV_gp3 large T antigen Mouse Monoclonal Antibody [clone: Ab3]
Katalogové číslo ABCAAB202866-100
Supplier: Abcam
Specifications
- Size:100 µg
- Pack type:Vial
Specifications
About this item
Mouse monoclonal [Ab3] to MCPyV_gp3 large T antigen.
Type: Primary
Antigen: MCPyV_gp3 large T antigen
Clonality: Monoclonal
Clone: Ab3
Conjugation:
Epitope: The epitope for monoclonal Ab3 is between amino acid residues 79 and 260 of MCPyV large T antigen. Ab3 does not bind to MCPyV small T antigen (PubMed ID: 23114601).
Host: Mouse
Isotype: IgG1
Reactivity: