Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: SP2]
ABCAAB239793-100
: Abcam
- Size:100 µg
- Pack type:Vial
Rabbit monoclonal [SP2] to Progesterone Receptor - BSA and Azide free.
Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: SP2
Conjugation:
Epitope: Amino acids 412-526
Host: Rabbit
Isotype: IgG
Reactivity: Human