Anti-MiTF Rabbit Monoclonal Antibody [clone: EPR26363-10]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR26363-10] to MiTF.
Type: Primary
Antigen: MiTF
Clonality: Monoclonal
Clone: EPR26363-10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together