Order Entry
Czech Republic
ContactUsLinkComponent
Anti-NFkB p100/NFKB2 (phospho S865) Rabbit Monoclonal Antibody [clone: EPR4686-66] (Alexa Fluor® 488)
Anti-NFkB p100/NFKB2 (phospho S865) Rabbit Monoclonal Antibody [clone: EPR4686-66] (Alexa Fluor® 488)
Catalog # ABCAAB214265-100
Supplier:  Abcam
undefined
Anti-NFkB p100/NFKB2 (phospho S865) Rabbit Monoclonal Antibody [clone: EPR4686-66] (Alexa Fluor® 488)
Catalog # ABCAAB214265-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 488 Rabbit monoclonal [EPR4686-66] to NFkB p100/NFKB2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NFkB p100/NFKB2
Clonality: Monoclonal
Clone: EPR4686-66
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human