Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
Catalog # ABCAAB218983-50
Supplier:  Abcam
undefined
Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
Catalog # ABCAAB218983-50
Supplier:  Abcam

Specifications

  • Size:
    50 µg
  • Pack type:
    Vial

Specifications

About this item

Rabbit polyclonal to Carbonic Anhydrase 12/CA12.

Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human