Anti-MAP1B Rabbit Monoclonal Antibody [clone: EPR27225-25]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR27225-25] to MAP1B.
Type: Primary
Antigen: MAP1B
Clonality: Monoclonal
Clone: EPR27225-25
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together