Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CEACAM5 Rabbit Monoclonal Antibody [clone: 61] (APC (Allophycocyanin))
Anti-CEACAM5 Rabbit Monoclonal Antibody [clone: 61] (APC (Allophycocyanin))
Supplier:  Abcam
Certificates
undefined
Anti-CEACAM5 Rabbit Monoclonal Antibody [clone: 61] (APC (Allophycocyanin))
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB275585-100
  • ABCAAB275585-25
  • Antigen symbol:
  • CEACAM5
  • CEACAM5
  • Antigen name:
  • CD66e
  • CD66e
  • Antigen synonyms:
  • CEA|Cell adhesion molecule CEACAM5|Carcinoembryonic antigen|Carcinoembryonic antigen-related cell adhesion molecule 5|Meconium antigen 100|CEA cell adhesion molecule 5
  • CEA|Cell adhesion molecule CEACAM5|Carcinoembryonic antigen|Carcinoembryonic antigen-related cell adhesion molecule 5|Meconium antigen 100|CEA cell adhesion molecule 5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 61
  • 61
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Full Length Protein corresponding to Human CEACAM5.
  • Recombinant Full Length Protein corresponding to Human CEACAM5.
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 76795 Da
  • 76795 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage temperature:
  • Store at 4 °C. Do not freeze. Store in the dark.
  • Store at 4 °C. Do not freeze. Store in the dark.
  • Shipping temperature:
  • 4 °C
  • 4 °C

Specifications

Frequently Bought Together