Order Entry
Czech Republic
ContactUsLinkComponent
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: AA2] (Alexa Fluor® 647)
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: AA2] (Alexa Fluor® 647)
Catalog # ABCAAB235759-100
Supplier:  Abcam
undefined
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: AA2] (Alexa Fluor® 647)
Catalog # ABCAAB235759-100
Supplier:  Abcam

Specifications

  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 647 Mouse monoclonal [AA2] to beta Tubulin.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: AA2
Conjugation: Alexa Fluor® 647
Epitope: Recognizes amino acids 416-431
Host: Mouse
Isotype: IgG1
Reactivity: Human