Order Entry
Czech Republic
ContactUsLinkComponent
Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC64] (Alexa Fluor® 647)
Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC64] (Alexa Fluor® 647)
Catalog # ABCAAB224025-100
Supplier:  Abcam
undefined
Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC64] (Alexa Fluor® 647)
Catalog # ABCAAB224025-100
Supplier:  Abcam

Specifications

  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 647 Rabbit monoclonal [mOC64] to beta Amyloid 1-42.

Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Monoclonal
Clone: mOC64
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human